accessibility menu, dialog, popup

UserName

CAS RN 154702-08-6

H3CCH3NHONOO

CAS RN 154702-08-6

IUPAC Name: N-(2-ethylhexyl)-4-nitrobenzamide
Synonyms: N-(2-ethylhexyl)-4-nitrobenzamide
Molecular Weight (g/mol): 278.35
Molecular Formula: C15H22N2O3
InChi Key: XYLYSNQGXATQBX-UHFFFAOYNA-N
SMILES: CCCCC(CC)CNC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results