accessibility menu, dialog, popup

UserName

CAS RN 1530-39-8

ClClP

CAS RN 1530-39-8

IUPAC Name: [(4-chlorophenyl)methyl]triphenylphosphanium chloride
Synonyms: [(4-chlorophenyl)methyl]triphenylphosphanium chloride
Molecular Weight (g/mol): 423.32
Molecular Formula: C25H21Cl2P
InChi Key: RAHOAHBOOHXRDY-UHFFFAOYSA-M
SMILES: [Cl-].ClC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results