CAS RN 152922-71-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(Hydroxymethyl)cyclopropaneacetonitrile (>85%), TRC
CAS: 152922-71-9 Molecular Formula: C6H9NO Molecular Weight (g/mol): 111.14 Synonym: 2-(1-(hydroxymethyl)cyclopropyl)acetonitrile,[1-(Hydroxymethyl)cyclopropyl]acetonitrile IUPAC Name: 2-[1-(hydroxymethyl)cyclopropyl]acetonitrile SMILES: OCC1(CC#N)CC1