CAS RN 149724-31-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(3-Indolylmethyl)-L-tryptophan, TRC
CAS: 149724-31-2 Molecular Formula: C20H19N3O2 Molecular Weight (g/mol): 333.39 Synonym: 2-(1H-Indol-3-ylmethyl)-L-tryptophan,rac-(R)-3-(2-((1H-Indol-3-yl)methyl)-1H-indol-3-yl)-2-aminopropanoic Acid IUPAC Name: (2S)-2-amino-3-[2-(1H-indol-3-ylmethyl)-1H-indol-3-yl]propanoic acid SMILES: N[C@@H](Cc1c(Cc2c[nH]c3ccccc23)[nH]c4ccccc14)C(=O)O