CAS RN 1489389-18-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CCT245737, MedChemExpress
MedChemExpress CCT245737 is an orally active and seletive Chk1 inhibitor, with an IC50 of 1.3 nM.