accessibility menu, dialog, popup

UserName

CH3CH3OCH3CH3OCH3CH3OEr

CAS RN 14814-07-4

IUPAC Name: erbium(3+) tris(propan-2-olate)
Synonyms: erbium(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 344.52
Molecular Formula: C9H21ErO3
InChi Key: VOCNVTAXVORJBI-UHFFFAOYSA-N
SMILES: [Er+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)