CAS RN 147795-39-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-3-oxo-octanoyl-L-Homoserine lactone, TRC
CAS: 147795-39-9 Molecular Formula: C12 H19 N O4 Molecular Weight (g/mol): 241.284 IUPAC Name: 3-oxo-N-[(3S)-2-oxooxolan-3-yl]octanamide SMILES: CCCCCC(=O)CC(=O)N[C@H]1CCOC1=O