accessibility menu, dialog, popup

UserName

OHN

CAS RN 14735-70-7

IUPAC Name: 3-azatricyclo[4.2.1.0²,⁵]non-7-en-4-one
Synonyms: 3-azatricyclo[4.2.1.0²,⁵]non-7-en-4-one
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: WBZQHVKGKQXWMW-UHFFFAOYNA-N
SMILES: O=C1NC2C1C1CC2C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)