accessibility menu, dialog, popup

UserName

OOCdOOH3CH3CCH3CH3

CAS RN 14689-45-3

IUPAC Name: bis(4-hydroxypent-3-en-2-one) cadmium
Synonyms: bis(4-hydroxypent-3-en-2-one) cadmium
Molecular Weight (g/mol): 312.65
Molecular Formula: C10H16CdO4
InChi Key: PFWWQOLXBCJYKI-UHFFFAOYSA-N
SMILES: [Cd].CC(O)=CC(C)=O.CC(O)=CC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results