CAS RN 145119-17-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2E)-6-[[(1,1-Dimethylethoxy)carbonyl]amino]-2-hexenoic Acid, TRC
CAS: 145119-17-1 Molecular Formula: C11H19NO4 Molecular Weight (g/mol): 229.27 Synonym: (E)-6-[[(1,1-Dimethylethoxy)carbonyl]amino]-2-hexenoic Acid IUPAC Name: (E)-6-((tert-butoxycarbonyl)amino)hex-2-enoic acid SMILES: OC(/C=C/CCCNC(OC(C)(C)C)=O)=O