CAS RN 1450-74-4
IUPAC Name:
1-(5-chloro-2-hydroxyphenyl)ethan-1-one
Synonyms:
1-(5-chloro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol):
170.59
Molecular Formula:
C8H7ClO2
InChi Key:
XTGCUDZCCIRWHL-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(Cl)=CC=C1O
1
–
1
of
1
results
1
5'-Chloro-2'-hydroxyacetophenone, 99%, Thermo Scientific Chemicals
CAS: 1450-74-4 Molecular Formula: C8H7ClO2 Molecular Weight (g/mol): 170.592 MDL Number: MFCD00067788 InChI Key: XTGCUDZCCIRWHL-UHFFFAOYSA-N Synonym: 5'-chloro-2'-hydroxyacetophenone,1-5-chloro-2-hydroxyphenyl ethanone,5-chloro-2-hydroxyacetophenone,1-5-chloro-2-hydroxyphenyl ethan-1-one,2-acetyl-4-chlorophenol,ethanone, 1-5-chloro-2-hydroxyphenyl,2'-hydroxy-5'-chloroacetophenone,acetophenone, 5'-chloro-2'-hydroxy,2-hydroxy-5-chloro acetophenone,1-5-chloro-2-hydroxyphenyl-1-ethanone PubChem CID: 74061 IUPAC Name: 1-(5-chloro-2-hydroxyphenyl)ethanone SMILES: CC(=O)C1=C(C=CC(=C1)Cl)O