accessibility menu, dialog, popup

UserName

BONCH3NOOFHOOH

CAS RN 1449131-87-6

IUPAC Name: {2-fluoro-5-[methyl(2-nitrophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-fluoro-5-[methyl(2-nitrophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 318.07
Molecular Formula: C14H12BFN2O5
InChi Key: UOLFNPGGUUSEKJ-UHFFFAOYSA-N
SMILES: CN(C(=O)C1=CC(B(O)O)=C(F)C=C1)C1=CC=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results