CAS RN 14476-38-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
8-Heptadecanone, TRC
CAS: 14476-38-1 Molecular Formula: C17H34O Molecular Weight (g/mol): 254.45 Synonym: Heptadecan-8-one IUPAC Name: heptadecan-8-one SMILES: CCCCCCCCCC(=O)CCCCCCC