CAS RN 14446-75-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
cis-3,5-Dimethylpiperidine, TRC
CAS: 14446-75-4 Molecular Formula: C7H15N Molecular Weight (g/mol): 113.2 SMILES: C[C@@H]1CNC[C@H](C)C1