accessibility menu, dialog, popup

UserName

CAS RN 14429-02-8

IUPAC Name: benzyl[(4-methoxyphenyl)methyl]amine
Synonyms: benzyl[(4-methoxyphenyl)methyl]amine
Molecular Weight (g/mol): 227.31
Molecular Formula: C15H17NO
InChi Key: WZUBICZIDUTTNJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results