accessibility menu, dialog, popup

UserName

ClHONH2OH

CAS RN 14426-21-2

IUPAC Name: bis(2-hydroxyethyl)azanium chloride
Synonyms: bis(2-hydroxyethyl)azanium chloride
Molecular Weight (g/mol): 141.6
Molecular Formula: C4H12ClNO2
InChi Key: VJLOFJZWUDZJBX-UHFFFAOYSA-N
SMILES: [Cl-].OCC[NH2+]CCO
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)