CAS RN 1439896-74-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-(2,2,2-Trifluoroethoxy)pyrimidine-4-carboxylic Acid, TRC
CAS: 1439896-74-8 Molecular Formula: C7H5F3N2O3 Molecular Weight (g/mol): 222.12 IUPAC Name: 6-(2,2,2-trifluoroethoxy)pyrimidine-4-carboxylic acid SMILES: OC(=O)c1cc(OCC(F)(F)F)ncn1