CAS RN 143540-05-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Di-tert-butyl 2-(Hydroxymethyl) Piperazine-1,4-dicarboxylate, TRC
CAS: 143540-05-0 Molecular Formula: C15H28N2O5 Molecular Weight (g/mol): 316.39 SMILES: CC(C)(C)OC(=O)N1CCN(C(CO)C1)C(=O)OC(C)(C)C