accessibility menu, dialog, popup

UserName

CAS RN 143090-18-0

H3CH3CCH3OONHN

CAS RN 143090-18-0

IUPAC Name: tert-butyl N-(4-cyanophenyl)carbamate
Synonyms: tert-butyl N-(4-cyanophenyl)carbamate
Molecular Weight (g/mol): 218.26
Molecular Formula: C12H14N2O2
InChi Key: CPIRTCVNDVQQLE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results