accessibility menu, dialog, popup

UserName

H3COOCH3OONi

CAS RN 142600-62-2

IUPAC Name: nickel(2+) bis(2-methoxyethan-1-olate)
Synonyms: nickel(2+) bis(2-methoxyethanolate)
Molecular Weight (g/mol): 208.87
Molecular Formula: C6H14NiO4
InChi Key: KNEAYFOSQJIPKZ-UHFFFAOYSA-N
SMILES: [Ni++].COCC[O-].COCC[O-]
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results