accessibility menu, dialog, popup

UserName

CAS RN 1421266-64-9

NOHNH2NHOH2N(E/Z)(E/Z)

CAS RN 1421266-64-9

IUPAC Name: N'-hydroxy-2-[(N'-hydroxycarbamimidoyl)methyl]benzene-1-carboximidamide
Synonyms: N'-hydroxy-2-[(N'-hydroxycarbamimidoyl)methyl]benzenecarboximidamide
Molecular Weight (g/mol): 208.22
Molecular Formula: C9H12N4O2
InChi Key: CTCKPOCZAHVUEJ-UHFFFAOYSA-N
SMILES: NC(CC1=CC=CC=C1C(N)=NO)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results