accessibility menu, dialog, popup

UserName

CAS RN 14191-95-8

OHN

CAS RN 14191-95-8

IUPAC Name: 2-(4-hydroxyphenyl)acetonitrile
Synonyms: PHBC
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: AYKYOOPFBCOXSL-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC#N)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)