CAS RN 1417985-27-3
CAS RN 1417985-27-3
IUPAC Name:
magnesium(2+) (3,5-difluorophenyl)methanide bromide
Synonyms:
magnesium(2+) (3,5-difluorophenyl)methanide bromide
Molecular Weight (g/mol):
231.32
Molecular Formula:
C7H5BrF2Mg
InChi Key:
QAXANYLHEOPEGM-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].[CH2-]C1=CC(F)=CC(F)=C1