accessibility menu, dialog, popup

UserName

ClClPtNN

CAS RN 14096-51-6

IUPAC Name: platinum(2+) ethane-1,2-diamine dichloride
Synonyms: platinum(2+) ethylenediamine dichloride
Molecular Weight (g/mol): 326.08
Molecular Formula: C2H8Cl2N2Pt
InChi Key: LMABILRJNNFCPG-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pt++].NCCN
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)