CAS RN 14086-92-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3,4,6-Tetraacetate 2-(Benzoylamino)-2-deoxy-D-glucopyranose, TRC
CAS: 14086-92-1 Molecular Formula: C21 H25 N O10 Molecular Weight (g/mol): 451.424 Synonym: 1,3,4,6-Tetra-O-acetyl-2-benzoylamino-2-deoxy-D-glucopyranoside IUPAC Name: [(2R,3S,4R,5R)-3,4,6-triacetyloxy-5-benzamidooxan-2-yl]methyl acetate SMILES: CC(=O)OC[C@H]1OC(OC(=O)C)[C@H](NC(=O)c2ccccc2)[C@@H](OC(=O)C)[C@@H]1OC(=O)C