accessibility menu, dialog, popup

UserName

RhPPCBrOPhPhPhPhPhPh

CAS RN 14056-79-2

IUPAC Name: λ¹-rhodium(1+) methanediidylidyneoxidanium bis(triphenylphosphane) hydrobromidyl
Synonyms: λ¹-rhodium(1+) methanediidylidyneoxidanium bis(triphenylphosphine) hydrobromidyl
Molecular Weight (g/mol): 735.4
Molecular Formula: C37H30BrOP2Rh
InChi Key: XWGOAWLZLYWHJM-UHFFFAOYSA-N
SMILES: [Br].[Rh+].[C--]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results