CAS RN 140-86-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,4-Dihydroxy-2-butanone, >85%, TRC
CAS: 140-86-3 Molecular Formula: C4H8O3 Molecular Weight (g/mol): 104.1 IUPAC Name: 1,4-dihydroxybutan-2-one SMILES: OCCC(=O)CO