CAS RN 139143-09-2
IUPAC Name:
1,3-bis(propan-2-yl)-1H-imidazol-3-ium chloride
Synonyms:
1,3-diisopropylimidazol-1-ium chloride
Molecular Weight (g/mol):
188.7
Molecular Formula:
C9H17ClN2
InChi Key:
DOFXKPAOJLLPII-UHFFFAOYSA-M
SMILES:
[Cl-].CC(C)N1C=C[N+](=C1)C(C)C
1
–
2
of
2
results
1
1,3-Diisopropylimidazolium chloride, 97+%
CAS: 139143-09-2 Molecular Formula: C9H17ClN2 Molecular Weight (g/mol): 188.70 MDL Number: MFCD03840577 InChI Key: DOFXKPAOJLLPII-UHFFFAOYSA-M Synonym: 1,3-diisopropylimidazolium chloride,1,3-di-i-propylimidazolium chloride,1,3-diisopropyl-1h-imidazol-3-ium chloride,1,3-diisopropylimidazol-1-ium chloride,diic,acmc-20ap45,n,n'-isopropyl imidazolium chloride,1h-imidazolium,1,3-bis 1-methylethyl-, chloride 1:1,n,n inverted exclamation marka-isopropyl imidazolium chloride PubChem CID: 11229242 IUPAC Name: 1,3-di(propan-2-yl)imidazol-1-ium;chloride SMILES: [Cl-].CC(C)N1C=C[N+](=C1)C(C)C