accessibility menu, dialog, popup

UserName

CAS RN 138689-79-9

IUPAC Name: 5-[4-(methylsulfanyl)phenyl]-2H-1,2,3,4-tetrazole
Synonyms: 5-[4-(methylsulfanyl)phenyl]-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 192.24
Molecular Formula: C8H8N4S
InChi Key: BKSCIIVDHYXBRU-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results