CAS RN 1383716-40-2
CAS RN 1383716-40-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Vps34-PIK-III, MedChemExpress
MedChemExpress Vps34-PIK-III is a potent and selective inhibitor of VPS34 with an IC50 of 18 nM.