CAS RN 13748-90-8
CAS RN 13748-90-8
IUPAC Name:
(2S)-2-hydroxy-4-methylpentanoic acid
Synonyms:
(+)-α-hydroxyisocaproate L-2-hydroxyisocaproic acid hydroxyisocaproic acid (S)-2-hydroxyisocaproic acid L-α-hydroxyisocaproate L-2-hydroxyisocaproate (S)-leucic acid (+)-2-hydroxyisocaproic acid L-a-hydroxyisocaproic acid L-leucic acid
Molecular Weight (g/mol):
132.16
Molecular Formula:
C6H12O3
InChi Key:
LVRFTAZAXQPQHI-YFKPBYRVSA-N
SMILES:
CC(C)C[C@H](O)C(O)=O
1
–
3
of
3
results
1
L-alpha-Hydroxyisocaproic acid, 98%
CAS: 13748-90-8 Molecular Formula: C6H12O3 Molecular Weight (g/mol): 132.16 InChI Key: LVRFTAZAXQPQHI-YFKPBYRVSA-N Synonym: l-leucic acid,s-2-hydroxy-4-methylpentanoic acid,2s-2-hydroxy-4-methylpentanoic acid,l-2-hydroxy-4-methylvaleric acid,s-leucic acid,s-2-hydroxy-4-methylvaleric acid,l-2-hydroxyisocaproic acid,l-alpha-hydroxyisocaproic acid,s---2-hydroxyisocaproic acid,pentanoic acid, 2-hydroxy-4-methyl-, 2s PubChem CID: 83697 ChEBI: CHEBI:44510 IUPAC Name: (2S)-2-hydroxy-4-methylpentanoic acid SMILES: CC(C)CC(C(=O)O)O
3