accessibility menu, dialog, popup

UserName

CH3OClFFFF

CAS RN 1373920-95-6

IUPAC Name: 4-fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride
Synonyms: 4-fluoro-3-methyl-5-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 240.58
Molecular Formula: C9H5ClF4O
InChi Key: CVJZTMWMSKRYDY-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC(=C1)C(Cl)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results