accessibility menu, dialog, popup

UserName

CAS RN 1373920-88-7

IUPAC Name: 4-fluoro-3-methyl-5-(trifluoromethyl)benzamide
Synonyms: 4-fluoro-3-methyl-5-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 221.16
Molecular Formula: C9H7F4NO
InChi Key: DBFJVYYVOCZDBU-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC(=C1)C(N)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results