accessibility menu, dialog, popup

UserName

NH2NBr

CAS RN 13534-98-0

IUPAC Name: 3-bromopyridin-4-amine
Synonyms: 4-amino-3-bromopyridine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: DDQYSZWFFXOXER-UHFFFAOYSA-N
SMILES: NC1=CC=NC=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)