CAS RN 1353000-19-7
IUPAC Name:
1-[3-fluoro-5-(trifluoromethoxy)phenyl]methanamine
Synonyms:
1-[3-fluoro-5-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol):
209.14
Molecular Formula:
C8H7F4NO
InChi Key:
ULLWGZMNUJLDKA-UHFFFAOYSA-N
SMILES:
NCC1=CC(OC(F)(F)F)=CC(F)=C1
1
–
1
of
1
results
1
3-Fluoro-5-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™
CAS: 1353000-19-7 Molecular Formula: C8H7F4NO Molecular Weight (g/mol): 209.144 MDL Number: MFCD18783230 InChI Key: ULLWGZMNUJLDKA-UHFFFAOYSA-N Synonym: 3-fluoro-5-trifluoromethoxy benzylamine,3-fluoro-5-trifluoromethoxy phenyl methanamine,1-3-fluoro-5-trifluoromethoxy phenyl methanamine PubChem CID: 66523552 IUPAC Name: [3-fluoro-5-(trifluoromethoxy)phenyl]methanamine SMILES: C1=C(C=C(C=C1OC(F)(F)F)F)CN