CAS RN 1352792-15-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3’-Hydroxy Repaglinide-d5(Mixture of Diastereomers), TRC
CAS: 1352792-15-4 Molecular Formula: C27H31D5N2O5 Molecular Weight (g/mol): 473.62 Synonym: 2-(Ethoxy-d5)-4-[2-[[1-[2-(3-hydroxy-1-piperidinyl)phenyl]-3-methylbutyl]amino]-2-oxoethyl]benzoic Acid IUPAC Name: 4-[2-[[1-[2-(3-hydroxypiperidin-1-yl)phenyl]-3-methylbutyl]amino]-2-oxoethyl]-2-(1,1,2,2,2-pentadeuterioethoxy)benzoic acid SMILES: [2H]C([2H])([2H])C([2H])([2H])Oc1cc(CC(=O)NC(CC(C)C)c2ccccc2N3CCCC(O)C3)ccc1C(=O)O