accessibility menu, dialog, popup

UserName

CAS RN 1347815-21-7

IUPAC Name: naphthalene-1,4-dicarbothioamide
Synonyms: naphthalene-1,4-dicarbothioamide
Molecular Weight (g/mol): 246.35
Molecular Formula: C12H10N2S2
InChi Key: RMVWFHNLLYPEGN-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(C(N)=S)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results