accessibility menu, dialog, popup

UserName

CAS RN 13472-79-2

INHO

CAS RN 13472-79-2

IUPAC Name: 5-iodo-1,2-dihydropyridin-2-one
Synonyms: 2(1H)-pyridinone, 5-iodo-
Molecular Weight (g/mol): 221
Molecular Formula: C5H4INO
InChi Key: ZDJUNNCVIDKJAN-UHFFFAOYSA-N
SMILES: IC1=CNC(=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)