CAS RN 1346603-20-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(1-Hydroxyethyl) Promazine-d4, TRC
CAS: 1346603-20-0 Molecular Formula: C19H20D4N2OS Molecular Weight (g/mol): 332.5 Synonym: 10-[3-(Dimethylamino)propyl]-α-methyl-10H-phenothiazine-2-methanol-d4 SMILES: [2H]C(O)(C([2H])([2H])[2H])C(C=C1)=CC2=C1SC3=CC=CC=C3N2CCCN(C)C