accessibility menu, dialog, popup

UserName

H2OFeOSOOO

CAS RN 13463-43-9

IUPAC Name: λ²-iron(2+) hydrate sulfate
Synonyms: λ²-iron(2+) hydrate sulfate
Molecular Weight (g/mol): 169.92
Molecular Formula: FeH2O5S
InChi Key: XBDUTCVQJHJTQZ-UHFFFAOYSA-L
SMILES: O.[Fe++].[O-]S([O-])(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results