accessibility menu, dialog, popup

UserName

CAS RN 134227-41-1

IUPAC Name: bis[(4-fluorophenyl)methyl]amine
Synonyms: bis[(4-fluorophenyl)methyl]amine
Molecular Weight (g/mol): 233.26
Molecular Formula: C14H13F2N
InChi Key: YKFNPVWWPPJNAQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNCC2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results