accessibility menu, dialog, popup

UserName

CAS RN 1338-24-5

CH3OHO

CAS RN 1338-24-5

IUPAC Name: 3-(3-ethylcyclopentyl)propanoic acid
Synonyms: 3-(3-ethylcyclopentyl)propanoic acid
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: WVRFSLWCFASCIS-UHFFFAOYNA-N
SMILES: CCC1CCC(CCC(O)=O)C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)