accessibility menu, dialog, popup

UserName

CAS RN 133332-49-7

IUPAC Name: [3-(cyclopentyloxy)-4-methoxyphenyl]methanol
Synonyms: [3-(cyclopentyloxy)-4-methoxyphenyl]methanol
Molecular Weight (g/mol): 222.28
Molecular Formula: C13H18O3
InChi Key: JIRHAGAOHOYLNO-UHFFFAOYSA-N
SMILES: COC1=C(OC2CCCC2)C=C(CO)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results