CAS RN 133229-11-5
IUPAC Name:
N-[4-(3-{[2-(3,4-dichlorophenyl)ethyl](methyl)amino}-2-hydroxypropoxy)phenyl]methanesulfonamide hydrochloride
Synonyms:
N-[4-(3-{[2-(3,4-dichlorophenyl)ethyl](methyl)amino}-2-hydroxypropoxy)phenyl]methanesulfonamide hydrochloride
Molecular Weight (g/mol):
483.83
Molecular Formula:
C19H25Cl3N2O4S
InChi Key:
TXOARFPCQOBODS-UHFFFAOYNA-N
SMILES:
Cl.CN(CCC1=CC(Cl)=C(Cl)C=C1)CC(O)COC1=CC=C(NS(C)(=O)=O)C=C1
1
–
1
of
1
results
1
AM-92016 hydrochloride, MedChemExpress
MedChemExpress AM-92016 hydrochloride is a specific blocker of rectifier potassium current (IK). AM-92016 hydrochloride delays rectifier potassium channel (IK), repolarizes the membrane thereby restricting the duration of the nerve impulse thereby restricting the duration of the nerve impulse.