CAS RN 13256-32-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3-Dimethyl-3-nitrosourea (Contains 25-35% water), TRC
CAS: 13256-32-1 Molecular Formula: C3H7N3O2 Molecular Weight (g/mol): 117.11 Synonym: N,N'-Dimethyl-N'-nitrosourea,1,3-Dimethyl-1-nitrosourea,1,3-Dimethylnitrosourea,1-Nitroso-1,3-dimethylurea,Dimethylnitrosourea,Dimethylnur,Dimethynur,N,N'-Dimethyl-N-nitrosourea,N,N'-Dimethylnitrosourea,N-Methyl-N'-methyl-N-nitrosourea,N-Nitroso-N,N'-dimethylurea,NDMU,NSC 48893,Nitrosodimethylurea SMILES: CNC(=O)N(C)N=O