accessibility menu, dialog, popup

UserName

CAS RN 1323966-29-5

CH3OFNCl

CAS RN 1323966-29-5

IUPAC Name: 2-(4-chloro-2-fluoro-3-methoxyphenyl)acetonitrile
Synonyms: 2-(4-chloro-2-fluoro-3-methoxyphenyl)acetonitrile
Molecular Weight (g/mol): 199.61
Molecular Formula: C9H7ClFNO
InChi Key: NXSFOSWMRVGGIA-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(CC#N)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results