CAS RN 1323966-18-2
IUPAC Name:
2-fluoro-5-methyl-4-(trifluoromethyl)benzoyl chloride
Synonyms:
2-fluoro-5-methyl-4-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol):
240.58
Molecular Formula:
C9H5ClF4O
InChi Key:
UDEWPGOZXPREEI-UHFFFAOYSA-N
SMILES:
CC1=C(C=C(F)C(=C1)C(Cl)=O)C(F)(F)F
1
–
1
of
1
results
1
2-Fluoro-5-methyl-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
CAS: 1323966-18-2 Molecular Formula: C9H5ClF4O Molecular Weight (g/mol): 240.582 MDL Number: MFCD19687193 InChI Key: UDEWPGOZXPREEI-UHFFFAOYSA-N Synonym: 2-fluoro-5-methyl-4-trifluoromethyl benzoyl chloride PubChem CID: 66523631 IUPAC Name: 2-fluoro-5-methyl-4-(trifluoromethyl)benzoyl chloride SMILES: CC1=C(C=C(C(=C1)C(=O)Cl)F)C(F)(F)F