accessibility menu, dialog, popup

UserName

CAS RN 1323966-13-7

IUPAC Name: 3-(4-chloro-2-fluoro-3-methoxyphenyl)propanoic acid
Synonyms: 3-(4-chloro-2-fluoro-3-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 232.64
Molecular Formula: C10H10ClFO3
InChi Key: MJIGCENLUSLAFF-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(CCC(O)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results