CAS RN 1322625-07-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-Hydroxyethyl)-1-deoxy-L-altronojirimycin Hydrochloride Salt, TRC
CAS: 1322625-07-9 IUPAC Name: (2S,3S,4R,5S)-1-(2-hydroxyethyl)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride