accessibility menu, dialog, popup

UserName

ClClH2NOH(R)NH2OHOH2NOH(S)NH2OHOHH

CAS RN 13204-98-3

IUPAC Name: dihydrogen (2R)-2,6-diamino-5-hydroxyhexanoic acid (2S)-2,6-diamino-5-hydroxyhexanoic acid dichloride
Synonyms: dihydrogen (2R)-2,6-diamino-5-hydroxyhexanoic acid 5-hydroxy-L-lysine dichloride
Molecular Weight (g/mol): 397.29
Molecular Formula: C12H30Cl2N4O6
InChi Key: NUMXTOLPCSNQQR-ZAKYRCQNSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCC(O)CC[C@H](N)C(O)=O.NCC(O)CC[C@@H](N)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)